Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-8
Entrez Gene ID
175660
Gene Name
Probable DNA-directed RNA polymerases I, II, and III subunit RPABC3
Gene Aliases
CELE_F26F4.11
Chromosome Location
Chr.III: 4917104 - 4918008 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065635.4 NP_498036.1 2 - 3 238 89

View Full Product Details