Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-4
Entrez Gene ID
175781
Gene Name
protein-coding
Gene Aliases
CELE_C05D2.1
Chromosome Location
Chr.III: 5625132 - 5632504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025977.6 NP_001021148.1 7 - 8 825 106
NM_001025978.3 NP_001021149.1 7 - 8 822 106
NM_001322590.3 NP_001309588.1 6 - 7 769 106
NM_001330884.3 NP_001317770.1 2 - 3 165 106
NM_065810.7 NP_498211.1 7 - 8 822 106

View Full Product Details