Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egg-1
Entrez Gene ID
175804
Gene Name
LDL receptor repeat-containing protein egg-1
Gene Aliases
CELE_B0244.8
Chromosome Location
Chr.III: 5728174 - 5730159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065836.10 NP_498237.2 2 - 3 150 71

View Full Product Details