Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-9
Entrez Gene ID
191835
Gene Name
Serpentine receptor class gamma-9
Gene Aliases
CELE_T12A2.10
Chromosome Location
Chr.III: 6255459 - 6256775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065964.1 NP_498365.1 2 - 3 770 82

View Full Product Details