Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-2
Entrez Gene ID
182793
Gene Name
Serpentine receptor class gamma-2
Gene Aliases
CELE_C18F10.5
Chromosome Location
Chr.III: 6258590 - 6260215 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065966.2 NP_498367.2 4 - 5 784 88

View Full Product Details