Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdfr-1
Entrez Gene ID
175942
Gene Name
protein-coding
Gene Aliases
CELE_C13B9.4
Chromosome Location
Chr.III: 6632837 - 6652083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025999.4 NP_001021170.1 5 - 6 872 117
NM_001026000.4 NP_001021171.1 5 - 6 872 117
NM_001306692.3 NP_001293621.1 3 - 4 235 117
NM_001372610.2 NP_001360187.1 3 - 4 229 117
NM_001382898.1 3 - 4 229 117
GenBank mRNA AY314776.1 - 5 - 6 872 117
AY314777.1 - 5 - 6 872 117
AY314778.1 - 5 - 6 872 117
EF141316.1 - 2 - 3 150 117
EF141317.1 - 2 - 3 150 117
EF141318.1 - 2 - 3 150 117

View Full Product Details