Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
isw-1
Entrez Gene ID
175944
Gene Name
Chromatin-remodeling complex ATPase chain isw-1
Gene Aliases
CELE_F37A4.8
Chromosome Location
Chr.III: 6703988 - 6710141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066067.6 NP_498468.2 8 - 9 2145 66

View Full Product Details