Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-1
Entrez Gene ID
191838
Gene Name
Serpentine receptor class V-1
Gene Aliases
CELE_R13F6.3
Chromosome Location
Chr.III: 6850385 - 6853203 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066088.7 NP_498489.2 3 - 4 559 153

View Full Product Details