Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-8
Entrez Gene ID
175981
Gene Name
Cuticle collagen 8
Gene Aliases
CELE_F11H8.3
Chromosome Location
Chr.III: 7019583 - 7020606 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382906.1 2 - 3 825 70

View Full Product Details