Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-2
Entrez Gene ID
175999
Gene Name
protein-coding
Gene Aliases
CELE_B0280.12
Chromosome Location
Chr.III: 7138640 - 7144959 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025942.4 NP_001021113.1 10 - 11 1425 73
NM_001025943.3 NP_001021114.1 12 - 13 1677 73
GenBank mRNA AF318606.2 - 12 - 13 1550 73

View Full Product Details