Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-21
Entrez Gene ID
176011
Gene Name
40S ribosomal protein S21
Gene Aliases
CELE_F37C12.11
Chromosome Location
Chr.III: 7186420 - 7189631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066178.9 NP_498579.2 2 - 3 145 77

View Full Product Details