Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmgr-1
Entrez Gene ID
176047
Gene Name
3-hydroxy-3-methylglutaryl coenzyme A reductase
Gene Aliases
CELE_F08F8.2
Chromosome Location
Chr.III: 7357561 - 7362132 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066225.7 NP_498626.2 8 - 9 1699 83

View Full Product Details