Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncr-2
Entrez Gene ID
176165
Gene Name
NPC intracellular cholesterol transporter 1 homolog 2
Gene Aliases
CELE_F09G8.4
Chromosome Location
Chr.III: 8258345 - 8265805 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066412.4 NP_498813.1 2 - 3 641 101

View Full Product Details