Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpn-3
Entrez Gene ID
176196
Gene Name
26S proteasome non-ATPase regulatory subunit 3
Gene Aliases
CELE_C30C11.2
Chromosome Location
Chr.III: 8447353 - 8449346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066468.5 NP_498869.1 1 - 2 114 68

View Full Product Details