Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-7
Entrez Gene ID
192073
Gene Name
TWiK family of potassium channels protein 7
Gene Aliases
CELE_F22B7.7
Chromosome Location
Chr.III: 8634067 - 8643739 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066502.6 NP_498903.3 3 - 4 313 71

View Full Product Details