Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F54F2.14
Entrez Gene ID
27219622
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F54F2.14
Chromosome Location
Chr.III: 8791545 - 8806286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379849.1 NP_001368592.1 14 - 15 3698 82

Gene Symbol
zfp-1
Entrez Gene ID
176237
Gene Name
protein-coding
Gene Aliases
CELE_F54F2.2
Chromosome Location
Chr.III: 8791545 - 8806289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382947.1 14 - 15 3795 82
NM_001382948.1 13 - 14 4235 82
NR_136383.1 13 - 14 2940 82

View Full Product Details