Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
F54F2.14
Entrez Gene ID
27219622
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F54F2.14
Chromosome Location
Chr.III: 8791545 - 8806286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379849.1 NP_001368592.1 11 - 12 1970 89

Gene Symbol
zfp-1
Entrez Gene ID
176237
Gene Name
protein-coding
Gene Aliases
CELE_F54F2.2
Chromosome Location
Chr.III: 8791545 - 8806289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382947.1 11 - 12 2067 89
NM_001382948.1 10 - 11 2507 89
NR_136383.1 10 - 11 1212 89

View Full Product Details