Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-11
Entrez Gene ID
176288
Gene Name
Protein ced-11
Gene Aliases
CELE_ZK512.3
Chromosome Location
Chr.III: 9119556 - 9125162 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066620.6 NP_499021.1 4 - 5 1590 123

View Full Product Details