Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dgk-3
Entrez Gene ID
186262
Gene Name
Probable diacylglycerol kinase 3
Gene Aliases
CELE_F54G8.2
Chromosome Location
Chr.III: 9163068 - 9168196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066630.4 NP_499031.3 4 - 5 528 126

View Full Product Details