Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
emb-9
Entrez Gene ID
176314
Gene Name
protein-coding
Gene Aliases
CELE_K04H4.1
Chromosome Location
Chr.III: 9336881 - 9344447 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027491.7 NP_001022662.2 11 - 12 5231 112
NM_001027492.6 NP_001022663.1 9 - 10 4411 112

View Full Product Details