The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_066673.7 | NP_499074.2 | 2 - 3 | 418 | 67 |
GenBank mRNA | AF192400.1 | - | 2 - 3 | 337 | 67 |