Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-6
Entrez Gene ID
182354
Gene Name
Putative sodium/calcium exchanger 6
Gene Aliases
CELE_C07A9.4
Chromosome Location
Chr.III: 9710240 - 9713949 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066745.3 NP_499146.2 4 - 5 723 159

View Full Product Details