Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyld-1
Entrez Gene ID
176401
Gene Name
protein-coding
Gene Aliases
CELE_F40F12.5
Chromosome Location
Chr.III: 9898575 - 9910658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268116.2 NP_001255045.1 3 - 4 443 62

View Full Product Details