Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-55
Entrez Gene ID
188561
Gene Name
Serpentine receptor class T-55
Gene Aliases
CELE_T16H12.8
Chromosome Location
Chr.III: 10104841 - 10106490 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066842.1 NP_499243.1 5 - 6 950 135

View Full Product Details