Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rde-4
Entrez Gene ID
176438
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T20G5.11
Chromosome Location
Chr.III: 10217302 - 10218646 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066864.5 NP_499265.1 2 - 3 611 111
GenBank mRNA AY071926.1 - 2 - 3 609 111

View Full Product Details