Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-69
Entrez Gene ID
176444
Gene Name
protein-coding
Gene Aliases
CELE_T07A5.6
Chromosome Location
Chr.III: 10318939 - 10320019 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027581.5 NP_001022752.1 1 - 2 195 80
NM_001027582.4 NP_001022753.1 1 - 2 195 80
NM_001129245.4 NP_001122717.1 1 - 2 132 80
GenBank mRNA AY919833.1 - 1 - 2 135 80

Gene Symbol
sas-7
Entrez Gene ID
176445
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T07C4.10
Chromosome Location
Chr.III: 10319006 - 10325675 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027583.4 NP_001022754.1 1 - 2 128 80

View Full Product Details