Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab-30
Entrez Gene ID
176476
Gene Name
RAB family
Gene Aliases
CELE_Y45F3A.2
Chromosome Location
Chr.III: 10560596 - 10565603 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066927.5 NP_499328.1 2 - 3 306 74

View Full Product Details