Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hpr-9
Entrez Gene ID
176483
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.23
Chromosome Location
Chr.III: 10621551 - 10626398 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379907.1 NP_001368495.1 10 - 11 1387 96
NM_066941.7 NP_499342.3 10 - 11 1465 96

View Full Product Details