Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hpl-2
Entrez Gene ID
176506
Gene Name
protein-coding
Gene Aliases
CELE_K01G5.2
Chromosome Location
Chr.III: 10752195 - 10755530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027481.5 NP_001022652.1 1 - 2 52 80
NM_001027482.4 NP_001022653.1 1 - 2 61 80
NM_001027483.3 NP_001022654.1 1 - 2 52 80
GenBank mRNA AF123573.1 - 1 - 2 46 80
AF123574.1 - 1 - 2 52 80

View Full Product Details