Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsa-2
Entrez Gene ID
176536
Gene Name
protein-coding
Gene Aliases
CELE_Y48A6B.11
Chromosome Location
Chr.III: 11063479 - 11068758 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027715.3 NP_001022886.1 4 - 5 873 120
NM_001027716.4 NP_001022887.1 4 - 5 802 120

View Full Product Details