Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cku-70
Entrez Gene ID
176546
Gene Name
Ku domain-containing protein
Gene Aliases
CELE_Y47D3A.4
Chromosome Location
Chr.III: 11154827 - 11168821 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067037.5 NP_499438.2 1 - 2 229 95

View Full Product Details