Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-8
Entrez Gene ID
189976
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_Y47D3A.11
Chromosome Location
Chr.III: 11218791 - 11222733 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067041.3 NP_499442.2 11 - 12 1495 58

View Full Product Details