Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crh-1
Entrez Gene ID
176597
Gene Name
protein-coding
Gene Aliases
CELE_Y41C4A.4
Chromosome Location
Chr.III: 11657738 - 11695165 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027688.5 NP_001022859.1 3 - 4 347 65
NM_001027690.3 NP_001022861.1 3 - 4 241 65
NM_001027691.4 NP_001022862.1 3 - 4 363 65
NM_001027692.4 NP_001022863.1 3 - 4 248 65
NM_001268206.3 NP_001255135.1 2 - 3 563 65
GenBank mRNA AB081595.1 - 3 - 4 248 65
AB081596.1 - 3 - 4 330 65

View Full Product Details