Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-8
Entrez Gene ID
176613
Gene Name
protein-coding
Gene Aliases
CELE_C18D11.4
Chromosome Location
Chr.III: 11826418 - 11831006 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268213.3 NP_001255142.1 1 - 2 60 75

View Full Product Details