Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trap-4
Entrez Gene ID
176626
Gene Name
TRanslocon-Associated Protein
Gene Aliases
CELE_Y56A3A.21
Chromosome Location
Chr.III: 11920735 - 11923827 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383006.1 2 - 3 385 71

View Full Product Details