Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrp-8
Entrez Gene ID
190710
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y75B8A.26
Chromosome Location
Chr.III: 12283133 - 12303216 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067197.4 NP_499598.2 9 - 10 1178 132

View Full Product Details