Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpt-6
Entrez Gene ID
176661
Gene Name
26S proteasome regulatory subunit 8
Gene Aliases
CELE_Y49E10.1
Chromosome Location
Chr.III: 12370456 - 12372625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067208.5 NP_499609.1 1 - 2 128 102

View Full Product Details