Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-80
Entrez Gene ID
176683
Gene Name
protein-coding
Gene Aliases
CELE_Y111B2A.14
Chromosome Location
Chr.III: 12626568 - 12651761 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268248.4 NP_001255177.1 20 - 21 4137 69
NM_001268249.2 NP_001255178.1 19 - 20 4054 69
NM_001312958.3 NP_001299887.1 20 - 21 4153 69
NM_001312959.3 NP_001299888.1 19 - 19 4056 69
NM_001312960.3 NP_001299889.1 17 - 18 3567 69
NM_001312961.3 NP_001299890.1 16 - 17 3477 69
NM_001312962.3 NP_001299891.1 17 - 18 3558 69
NM_001312963.3 NP_001299892.1 16 - 17 3468 69
NM_001312964.3 NP_001299893.1 16 - 17 2997 69
NM_001312965.3 NP_001299894.1 15 - 16 2907 69
NM_001312966.3 NP_001299895.1 16 - 17 2988 69
NM_001312967.3 NP_001299896.1 15 - 16 2898 69
NM_001312968.3 NP_001299897.1 7 - 8 1569 69
NM_001312969.3 NP_001299898.1 6 - 7 1479 69
NM_001312970.3 NP_001299899.1 5 - 6 960 69
NM_001379963.1 NP_001368536.1 2 - 3 315 69
NM_001379964.1 NP_001368537.1 1 - 2 132 69

View Full Product Details