Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tpst-1
Entrez Gene ID
176684
Gene Name
Protein-tyrosine sulfotransferase A
Gene Aliases
CELE_Y111B2A.15
Chromosome Location
Chr.III: 12652340 - 12662286 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067245.5 NP_499646.3 4 - 5 1082 123
GenBank mRNA AF049709.1 - 4 - 5 1065 123

View Full Product Details