Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-37
Entrez Gene ID
176694
Gene Name
Ligand-Gated ion Channel
Gene Aliases
CELE_ZC482.5
Chromosome Location
Chr.III: 12785154 - 12793176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067261.7 NP_499662.2 1 - 2 87 103

View Full Product Details