Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drr-2
Entrez Gene ID
259546
Gene Name
RRM domain-containing protein
Gene Aliases
CELE_T12D8.2
Chromosome Location
Chr.III: 13642420 - 13643570 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067417.4 NP_499818.1 2 - 3 142 116

View Full Product Details