Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
toca-2
Entrez Gene ID
176812
Gene Name
protein-coding
Gene Aliases
CELE_K08E3.3
Chromosome Location
Chr.III: 13755896 - 13759445 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067437.6 NP_499838.2 14 - 15 1826 65

View Full Product Details