Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-44
Entrez Gene ID
191978
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C42D4.4
Chromosome Location
Chr.IV: 7177187 - 7178423 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068730.1 NP_501131.1 2 - 3 557 146

View Full Product Details