Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
valv-1
Entrez Gene ID
182010
Gene Name
LIM zinc-binding domain-containing protein
Gene Aliases
CELE_B0496.7
Chromosome Location
Chr.IV: 7444639 - 7445432 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068786.6 NP_501187.1 2 - 3 264 70

View Full Product Details