Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rig-4
Entrez Gene ID
177597
Gene Name
Protein sidekick homolog
Gene Aliases
CELE_Y42H9B.2
Chromosome Location
Chr.IV: 7991410 - 8004136 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068938.8 NP_501339.2 1 - 2 228 68

View Full Product Details