Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
praf-3
Entrez Gene ID
177637
Gene Name
protein-coding
Gene Aliases
CELE_D2096.2
Chromosome Location
Chr.IV: 8379983 - 8381936 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027933.3 NP_001023104.1 2 - 3 322 80

View Full Product Details