Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-8
Entrez Gene ID
177652
Gene Name
SaPosin-like Protein family
Gene Aliases
CELE_C28C12.5
Chromosome Location
Chr.IV: 8480846 - 8483710 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069046.6 NP_501447.1 7 - 8 1039 81

View Full Product Details