Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-77
Entrez Gene ID
177656
Gene Name
protein-coding
Gene Aliases
CELE_C28C12.10
Chromosome Location
Chr.IV: 8503492 - 8510548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307115.4 NP_001294044.2 5 - 6 932 88
NM_001307116.3 NP_001294045.1 3 - 4 580 88
NM_001307117.3 NP_001294046.1 3 - 4 580 88
NM_001380369.1 NP_001368458.1 3 - 4 382 88
NM_001380370.1 NP_001368457.1 3 - 4 382 88
NM_069054.6 NP_501455.2 5 - 6 933 88

View Full Product Details