Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dgk-4
Entrez Gene ID
177667
Gene Name
protein-coding
Gene Aliases
CELE_F42A9.1
Chromosome Location
Chr.IV: 8631333 - 8644019 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069075.6 NP_501476.2 13 - 14 2492 85

View Full Product Details