Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdh-5
Entrez Gene ID
177671
Gene Name
protein-coding
Gene Aliases
CELE_F08B4.2
Chromosome Location
Chr.IV: 8668935 - 8676619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307097.4 NP_001294026.1 15 - 16 2968 70
NM_001356929.4 NP_001343640.1 17 - 17 3400 70
NM_001356930.4 NP_001343703.1 15 - 15 3193 70
NM_069086.6 NP_501487.1 15 - 16 2889 70

View Full Product Details